siRNA Details
Gene Symbol: Tex37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028825.3 | NP_083101.1 | 3 | 153 | ||
GenBank mRNA | AK005862.1 | - | 3 | 154 | ||
BC049736.1 | - | 3 | 134 | 6771350 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
