siRNA Details
Gene Symbol: NACA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113201.2 | NP_001106672.1 | 7 | 953 | ||
NM_001113202.1 | NP_001106673.1 | 7 | 876 | |||
NM_001113203.2 | NP_001106674.2 | 10 | 3006 | |||
NM_001320193.1 | NP_001307122.1 | 7 | 902 | |||
NM_001320194.1 | NP_001307123.1 | 7 | 876 | |||
NM_005594.5 | NP_005585.1 | 7 | 927 | |||
XM_006719412.1 | XP_006719475.1 | 8 | 7224 | |||
XM_006719413.2 | XP_006719476.1 | 8 | 6480 | |||
XM_006719414.1 | XP_006719477.1 | 8 | 6222 | |||
XM_006719418.1 | XP_006719481.1 | 7 | 659 | |||
XM_006719420.3 | XP_006719483.1 | 7 | 915 | |||
XM_011538389.1 | XP_011536691.1 | 8 | 6222 | |||
XM_017019337.1 | XP_016874826.1 | 9 | 2796 | |||
GenBank mRNA | AF054187.1 | - | 7 | 903 | ||
AK090650.1 | - | 7 | 2533 | |||
AK096699.1 | - | 10 | 2763 | |||
AK223085.1 | - | 7 | 633 | |||
AK293209.1 | - | 7 | 495 | |||
AK311904.1 | - | 7 | 622 | |||
AW021139.1 | - | 3 | 266 | |||
AY034001.1 | - | 6 | 597 | |||
AY911673.1 | - | 7 | 619 | |||
BC105120.1 | - | 7 | 794 | |||
BC105122.1 | - | 7 | 794 | |||
BC106041.1 | - | 7 | 917 | |||
CD104788.1 | - | 7 | 649 | |||
CR450295.1 | - | 6 | 594 | |||
X80909.1 | - | 7 | 619 |
Gene Symbol: NACAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002182.1 | 1 | 607 | |||
GenBank mRNA | AF315951.1 | - | 2 | 669 | ||
AK291701.1 | - | 3 | 1139 | |||
BC069062.1 | - | 1 | 607 | |||
BC069411.1 | - | 1 | 700 | |||
BC074958.2 | - | 1 | 695 | |||
BC074959.2 | - | 1 | 695 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
