siRNA Details
Gene Symbol: NACA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113201.2 | NP_001106672.1 | 5 | 718 | ||
NM_001113202.1 | NP_001106673.1 | 5 | 641 | |||
NM_001113203.2 | NP_001106674.2 | 8 | 2771 | |||
NM_001320193.1 | NP_001307122.1 | 5 | 667 | |||
NM_001320194.1 | NP_001307123.1 | 5 | 641 | |||
NM_005594.5 | NP_005585.1 | 5 | 692 | |||
XM_006719412.1 | XP_006719475.1 | 6 | 6989 | |||
XM_006719413.2 | XP_006719476.1 | 6 | 6245 | |||
XM_006719414.1 | XP_006719477.1 | 6 | 5987 | |||
XM_006719418.1 | XP_006719481.1 | 5 | 424 | |||
XM_006719420.3 | XP_006719483.1 | 5 | 680 | |||
XM_011538389.1 | XP_011536691.1 | 6 | 5987 | |||
XM_017019337.1 | XP_016874826.1 | 7 | 2561 | |||
GenBank mRNA | AF054187.1 | - | 5 | 668 | ||
AK090650.1 | - | 5 | 2298 | |||
AK096699.1 | - | 8 | 2528 | |||
AK223085.1 | - | 5 | 398 | |||
AK293209.1 | - | 5 | 260 | |||
AK311904.1 | - | 5 | 387 | |||
AW021139.1 | - | 1 | 31 | |||
AY034001.1 | - | 4 | 362 | |||
AY605660.1 | - | 5 | 397 | |||
AY911673.1 | - | 5 | 384 | |||
BC105120.1 | - | 5 | 559 | |||
BC105122.1 | - | 5 | 559 | |||
BC106041.1 | - | 5 | 682 | |||
BI856347.1 | - | 5 | 465 | |||
CD104788.1 | - | 5 | 414 | |||
CD246364.1 | - | 5 | 656 | |||
CR450295.1 | - | 4 | 359 | |||
X80909.1 | - | 5 | 384 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
