siRNA Details
Gene Symbol: 1700016K19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198637.2 | NP_941039.1 | 2 | 489 | ||
GenBank mRNA | AK132806.1 | - | 2 | 414 | ||
BC049758.1 | - | 2 | 428 | 6773542 | ||
BQ931096.1 | - | 2 | 504 |

More Information
Other Species Targeted

Gene Ontology Categories:
