siRNA Details
Gene Symbol: NACA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF054187.1 | - | 4 | 484 | ||
AK090650.1 | - | 4 | 2114 | |||
AK096699.1 | - | 7 | 2344 | |||
AK293209.1 | - | 4 | 214 | |||
AK311904.1 | - | 4 | 203 | |||
AY034001.1 | - | 3 | 178 | |||
AY605660.1 | - | 4 | 213 | |||
BC105120.1 | - | 4 | 375 | |||
BC105122.1 | - | 4 | 375 | |||
BC106041.1 | - | 4 | 498 | |||
BG709003.1 | - | 4 | 502 | |||
CD104788.1 | - | 4 | 230 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
