siRNA Details
Gene Symbol: Smu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021535.4 | NP_067510.3 | 3, 4 | 482 | ||
GenBank mRNA | AB044414.1 | - | 3, 4 | 411 | ||
AK003493.1 | - | 3, 4 | 469 | |||
AK011140.1 | - | 3, 4 | 481 | |||
AK019174.1 | - | 3, 4 | 441 | |||
AK145982.1 | - | 3, 4 | 481 | |||
AK159635.1 | - | 3, 4 | 469 | |||
AK168813.1 | - | 3, 4 | 476 | |||
BC020034.1 | - | 3, 4 | 424 | |||
BC057446.1 | - | 3, 4 | 443 | 3155760 |
