siRNA Details
Gene Symbol: Chic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006962.1 | - | 4 | 703 | ||
AK172289.1 | - | 4 | 696 | |||
BC048435.1 | - | 5 | 456 | |||
BC116971.1 | - | 4 | 513 | 40126290 | ||
BC116973.1 | - | 4 | 513 | 40126292 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
