siRNA Details
Gene Symbol: 1700080O16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC109337.2 | - | 2, 3 | 191 | 40052428 | |
BC109338.1 | - | 1, 2 | 74 | 40052437 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
