siRNA Details
Gene Symbol: Spatc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028852.1 | NP_083128.1 | 2 | 436 | ||
GenBank mRNA | AB032199.1 | - | 2 | 436 | ||
AK006994.1 | - | 2 | 437 | |||
BC116725.1 | - | 2 | 403 | |||
BC118509.1 | - | 2 | 396 | 40106461 |
