siRNA Details
Gene Symbol: Kcmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019715.2 | NP_062689.2 | 2 | 427 | ||
GenBank mRNA | AK007070.1 | - | 2 | 374 | ||
AK142436.1 | - | 2 | 184 | |||
BC029325.1 | - | 2 | 249 | |||
BC043330.1 | - | 2 | 208 | 5249829 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
