siRNA Details
Gene Symbol: Mtmr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028860.2 | NP_083136.2 | 9, 10 | 1179 | ||
GenBank mRNA | AK141138.1 | - | 9, 10 | 1159 | ||
BC032166.1 | - | 9, 10 | 1162 | 5352919 | ||
BC081544.1 | - | 9, 10 | 1163 | 3672099 | ||
CV557784.1 | - | 2, 3 | 307 |
