siRNA Details
Gene Symbol: 1700092M07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177347.1 | NP_001170818.1 | 2 | 157 | ||
GenBank mRNA | AK007043.1 | - | 2 | 120 | ||
BC048571.1 | - | 1 | 241 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
