siRNA Details
Gene Symbol: Osbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152818.2 | NP_690031.2 | 7 | 1525 | ||
GenBank mRNA | AK007088.1 | - | 3 | 106 | ||
AK044598.1 | - | 4 | 313 | |||
BC031794.1 | - | 5 | 488 | 4501119 | ||
BC051185.1 | - | 5 | 484 | |||
BC058356.1 | - | 6 | 668 | 5696357 | ||
BC058602.1 | - | 5 | 488 | 6467653 | ||
CV784397.1 | - | 5 | 517 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
