siRNA Details
Gene Symbol: Cxxc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028868.3 | NP_083144.1 | 9 | 1347 | ||
GenBank mRNA | AK010337.1 | - | 9 | 1348 | ||
AK083655.1 | - | 9 | 1352 | |||
AK146688.1 | - | 9 | 1431 | |||
AY099096.1 | - | 9 | 1319 | |||
BC012261.1 | - | 5 | 890 | |||
BC030938.1 | - | 9 | 1179 | 3980339 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
