siRNA Details
Gene Symbol: Ranbp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145824.4 | NP_665823.2 | 13 | 1805 | ||
GenBank mRNA | AK087165.1 | - | 3 | 222 | ||
AK173177.1 | - | 13 | 1618 | |||
AY337314.1 | - | 13 | 1806 | |||
BC024698.1 | - | 13 | 1650 | 3667000 | ||
EU281316.1 | - | 13 | 1666 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
