siRNA Details
Gene Symbol: Slc6a19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028878.3 | NP_083154.1 | 6, 7 | 943 | ||
GenBank mRNA | AJ633679.1 | - | 6, 7 | 952 | ||
AK014544.1 | - | 6, 7 | 944 | |||
AK028784.1 | - | 6, 7 | 944 | |||
AK128923.1 | - | 5, 6 | 1286 | |||
AK162399.1 | - | 5, 6 | 688 | |||
BC107411.1 | - | 6, 7 | 944 | 40059042 | ||
BC107412.2 | - | 6, 7 | 948 | 40059050 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
