siRNA Details
Gene Symbol: Ahcyl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171000.2 | NP_001164471.2 | 11 | 1447 | ||
NM_001171001.1 | NP_001164472.1 | 11 | 1167 | |||
NM_021414.6 | NP_067389.5 | 11 | 1450 | |||
GenBank mRNA | AK014539.1 | - | 11 | 1398 | ||
AK053527.1 | - | 11 | 1131 | |||
AK122382.1 | - | 9 | 1153 | |||
AK148660.1 | - | 1 | 1122 | |||
BC051504.1 | - | 1 | 80 | |||
BC059255.1 | - | 7 | 622 | |||
BC079660.1 | - | 11 | 1360 | 6853550 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
