siRNA Details
Gene Symbol: Ahcyl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171000.2 | NP_001164471.2 | 8 | 1222 | ||
NM_001171001.1 | NP_001164472.1 | 8 | 942 | |||
NM_021414.6 | NP_067389.5 | 8 | 1225 | |||
GenBank mRNA | AK014539.1 | - | 8 | 1173 | ||
AK053527.1 | - | 8 | 906 | |||
AK122382.1 | - | 6 | 928 | |||
BC059255.1 | - | 4 | 397 | |||
BC079660.1 | - | 8 | 1135 | 6853550 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
