siRNA Details
Gene Symbol: Fam160a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242363.2 | NP_001229292.1 | 4 | 857 | ||
NM_001242364.2 | NP_001229293.1 | 3 | 766 | |||
NM_001242365.2 | NP_001229294.1 | 3 | 766 | |||
NM_199009.3 | NP_950174.1 | 3 | 766 | |||
GenBank mRNA | AK085350.1 | - | 3 | 715 | ||
AK122552.1 | - | 3 | 694 | |||
AK154173.1 | - | 4 | 801 | |||
AK155273.1 | - | 4 | 802 | |||
AK159716.1 | - | 3 | 721 | |||
AK169996.1 | - | 4 | 835 | |||
BC018464.1 | - | 3 | 696 | |||
BC021532.1 | - | 1 | 93 | |||
BC043717.1 | - | 2 | 282 | |||
BC063093.1 | - | 3 | 723 | 6332038 |
