siRNA Details
Gene Symbol: Fam160a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242363.2 | NP_001229292.1 | 4 | 682 | ||
NM_001242364.2 | NP_001229293.1 | 3 | 591 | |||
NM_001242365.2 | NP_001229294.1 | 3 | 591 | |||
NM_199009.3 | NP_950174.1 | 3 | 591 | |||
GenBank mRNA | AK085350.1 | - | 3 | 540 | ||
AK122552.1 | - | 3 | 519 | |||
AK154173.1 | - | 4 | 626 | |||
AK155273.1 | - | 4 | 627 | |||
AK159716.1 | - | 3 | 546 | |||
AK169996.1 | - | 4 | 659 | |||
BC018464.1 | - | 3 | 521 | |||
BC043717.1 | - | 2 | 107 | |||
BC063093.1 | - | 3 | 548 | 6332038 |

More Information
Other Species Targeted

Gene Ontology Categories:
