siRNA Details
Gene Symbol: 4931414P19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028890.2 | NP_083166.1 | 6 | 1479 | ||
GenBank mRNA | AK016457.1 | - | 5 | 952 | ||
AK163117.1 | - | 6 | 1488 | |||
BC029624.1 | - | 6 | 1479 | 3992239 | ||
BC034212.1 | - | 6 | 1480 |
