siRNA Details
Gene Symbol: 4931414P19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028890.2 | NP_083166.1 | 4, 5 | 1332 | ||
GenBank mRNA | AK016457.1 | - | 3, 4 | 805 | ||
AK163117.1 | - | 4, 5 | 1341 | |||
BC029624.1 | - | 4, 5 | 1332 | 3992239 | ||
BC034212.1 | - | 4, 5 | 1332 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
