siRNA Details
Gene Symbol: Dpp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028906.2 | NP_083182.2 | 17, 18 | 2443 | ||
GenBank mRNA | AK016546.1 | - | 17, 18 | 2482 | ||
AK029788.1 | - | 15, 16 | 4314 | |||
AK155605.1 | - | 17, 18 | 2490 | |||
AK171895.1 | - | 4, 5 | 513 | |||
BC043124.1 | - | 17, 18 | 2443 | 6410075 | ||
BC059222.1 | - | 17, 18 | 2443 | 6410075 |
