siRNA Details
Gene Symbol: Map10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028908.3 | NP_083184.1 | 1 | 1725 | ||
GenBank mRNA | AK016632.1 | - | 1 | 1681 | ||
AK042358.1 | - | 1 | 1698 | |||
AK080043.1 | - | 1 | 1679 | |||
AK220388.1 | - | 1 | 1652 |
