siRNA Details
Gene Symbol: Tmc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105252.1 | NP_001098722.1 | 8 | 1650 | ||
NM_028930.3 | NP_083206.1 | 4 | 801 | |||
GenBank mRNA | AK016573.1 | - | 4 | 778 | ||
AK028404.1 | - | 9 | 1782 | |||
AY236495.1 | - | 4 | 692 | |||
AY263159.1 | - | 4 | 801 | |||
BC046390.1 | - | 2 | 108 | |||
BC108382.1 | - | 7 | 1311 | 4457529 |

More Information
Other Species Targeted

Gene Ontology Categories:
