siRNA Details
Gene Symbol: Tmc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105252.1 | NP_001098722.1 | 11 | 2184 | ||
NM_028930.3 | NP_083206.1 | 7 | 1335 | |||
GenBank mRNA | AK016573.1 | - | 7 | 1312 | ||
AK028404.1 | - | 12 | 2316 | |||
AY236495.1 | - | 7 | 1226 | |||
AY263159.1 | - | 7 | 1335 | |||
BC046390.1 | - | 5 | 642 | |||
BC108382.1 | - | 10 | 1845 | 4457529 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
