siRNA Details
Gene Symbol: Arl13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028947.1 | NP_083223.1 | 6 | 785 | ||
GenBank mRNA | AK016872.1 | - | 6 | 786 | ||
BC100522.1 | - | 6 | 683 | 30920038 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
