siRNA Details
Gene Symbol: Pgs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133757.2 | NP_598518.1 | 7 | 1105 | ||
GenBank mRNA | AK016901.1 | - | 5 | 924 | ||
AK046174.1 | - | 7 | 1097 | |||
AK049359.1 | - | 7 | 1106 | |||
AK053905.1 | - | 7 | 1091 | |||
AK153001.1 | - | 5 | 798 | |||
AK163967.1 | - | 7 | 1079 | |||
AK177638.1 | - | 1 | 142 | |||
BC022918.1 | - | 2 | 227 | |||
BC151089.1 | - | 7 | 1205 | 9088002 | ||
BC151185.1 | - | 7 | 1205 | 9088074 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
