siRNA Details
Gene Symbol: Nsun6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165941.1 | NP_001159413.1 | 6 | 945 | ||
NM_001165942.1 | NP_001159414.1 | 4 | 722 | |||
NM_001165943.1 | NP_001159415.1 | 5 | 649 | |||
NM_028950.4 | NP_083226.3 | 5 | 832 | |||
GenBank mRNA | AK016629.1 | - | 5 | 650 | ||
AK016811.1 | - | 6 | 958 | |||
AK042750.1 | - | 4 | 722 | |||
AK077778.1 | - | 5 | 716 | |||
AK132428.1 | - | 5 | 799 | |||
BC025622.1 | - | 2 | 175 | |||
BC053751.1 | - | 5 | 800 | 6335896 |
