siRNA Details
Gene Symbol: Nsun6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165941.1 | NP_001159413.1 | 10 | 1442 | ||
NM_001165943.1 | NP_001159415.1 | 9 | 1146 | |||
NM_028950.4 | NP_083226.3 | 9 | 1329 | |||
GenBank mRNA | AK016629.1 | - | 9 | 1146 | ||
AK016811.1 | - | 10 | 1455 | |||
AK077778.1 | - | 9 | 1213 | |||
AK132428.1 | - | 9 | 1296 | |||
BC025622.1 | - | 6 | 672 | |||
BC053751.1 | - | 9 | 1297 | 6335896 |
