siRNA Details
Gene Symbol: 4933433C11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028961.1 | NP_083237.1 | 5 | 724 | ||
GenBank mRNA | AK017033.1 | - | 4 | 814 | ||
AK029738.1 | - | 5 | 682 | |||
BC050147.1 | - | 1 | 271 | |||
BC132286.1 | - | 5 | 724 | 40130563 | ||
BC138006.1 | - | 5 | 724 | 8861025 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
