siRNA Details
Gene Symbol: 4933427D14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028963.2 | NP_083239.1 | 3 | 903 | ||
GenBank mRNA | AK016946.1 | - | 3 | 792 | ||
AK032379.1 | - | 3 | 797 | |||
AK041396.1 | - | 3 | 784 | |||
AK173018.1 | - | 3 | 772 | |||
BC043106.1 | - | 3 | 903 | 6406803 | ||
BC059269.1 | - | 3 | 989 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
