siRNA Details
Gene Symbol: 4933427D14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028963.2 | NP_083239.1 | 2, 3 | 327 | ||
GenBank mRNA | AK016946.1 | - | 2, 3 | 216 | ||
AK032379.1 | - | 2, 3 | 221 | |||
AK041396.1 | - | 2, 3 | 209 | |||
AK043241.1 | - | 2, 3 | 386 | |||
AK173018.1 | - | 2, 3 | 196 | |||
BC043106.1 | - | 2, 3 | 327 | 6406803 | ||
BC059269.1 | - | 2, 3 | 413 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
