siRNA Details
Gene Symbol: Snx29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290148.1 | NP_001277077.1 | 14 | 1371 | ||
NM_028964.4 | NP_083240.2 | 21 | 2420 | |||
GenBank mRNA | AF399755.1 | - | 14 | 1326 | ||
AK017102.1 | - | 14 | 1372 |
