siRNA Details
Gene Symbol: Osbpl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148958.2 | NP_683761.1 | 8 | 1467 | ||
GenBank mRNA | AK017034.1 | - | 10 | 1798 | ||
BC027256.1 | - | 8 | 1467 | 2650570 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
