siRNA Details
Gene Symbol: Kbtbd13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028974.1 | NP_083250.1 | 1 | 1052 | ||
GenBank mRNA | AK017385.1 | - | 1 | 65 | ||
AK048611.1 | - | 1 | 945 | |||
BC139793.1 | - | 2 | 932 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
