siRNA Details
Gene Symbol: Neto2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081324.1 | NP_001074793.1 | 6 | 738 | ||
GenBank mRNA | AK017458.1 | - | 6 | 813 | ||
AK081036.1 | - | 1 | 403 | |||
AK081462.1 | - | 6 | 837 | |||
AK083512.1 | - | 6 | 868 | |||
AY138991.1 | - | 6 | 702 | |||
BC068168.1 | - | 6 | 738 | |||
BC151050.1 | - | 6 | 698 | 9087963 | ||
BC157941.1 | - | 6 | 677 | 9088121 | ||
BC172104.1 | - | 6 | 677 | 9054390 | ||
BC172139.1 | - | 6 | 677 | 9054809 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
