siRNA Details
Gene Symbol: Neto2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081324.1 | NP_001074793.1 | 9 | 1484 | ||
GenBank mRNA | AK017458.1 | - | 9 | 1561 | ||
AK081462.1 | - | 9 | 1583 | |||
AK083512.1 | - | 9 | 1614 | |||
AY138991.1 | - | 9 | 1448 | |||
BC068168.1 | - | 10 | 1660 | |||
BC151050.1 | - | 9 | 1444 | 9087963 | ||
BC157941.1 | - | 9 | 1423 | 9088121 | ||
BC172104.1 | - | 9 | 1423 | 9054390 | ||
BC172139.1 | - | 9 | 1423 | 9054809 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
