siRNA Details
Gene Symbol: 8430419L09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028982.4 | NP_083258.2 | 2 | 198 | ||
GenBank mRNA | AK018428.1 | - | 2 | 142 | ||
AK039401.1 | - | 2 | 134 | |||
AK155128.1 | - | 2 | 142 | |||
AK169892.1 | - | 2 | 199 | |||
BC079676.1 | - | 2 | 125 | |||
BC137676.1 | - | 2 | 177 | 8860695 |
