siRNA Details
Gene Symbol: Ocstamp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029021.1 | NP_083297.1 | 2 | 819 | ||
GenBank mRNA | AK014751.1 | - | 2 | 819 | ||
AK032330.1 | - | 214 | ||||
AK042271.1 | - | 2 | 831 | |||
BC020160.1 | - | 2 | 787 | 3966086 |
