siRNA Details
Gene Symbol: S100pbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029036.2 | NP_083312.1 | 3 | 653 | ||
GenBank mRNA | AK015229.1 | - | 3 | 653 | ||
AK139097.1 | - | 3 | 627 | |||
AK146310.1 | - | 3 | 716 | |||
AK169364.1 | - | 3 | 618 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
