siRNA Details
Gene Symbol: Spdyb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029048.3 | NP_083324.3 | 7 | 1621 | ||
GenBank mRNA | AK015441.1 | - | 7 | 1622 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
