siRNA Details
Gene Symbol: Ccdc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197041.1 | NP_001183970.1 | 3 | 572 | ||
NM_029061.3 | NP_083337.1 | 3 | 572 | |||
GenBank mRNA | AK015811.1 | - | 3 | 554 | ||
AK016006.1 | - | 3 | 572 | |||
AY854152.1 | - | 3 | 604 | |||
EF100607.1 | - | 3 | 572 |
