siRNA Details
Gene Symbol: Pih1d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029062.2 | NP_083338.1 | 1 | 223 | ||
GenBank mRNA | AK015858.1 | - | 1 | 224 | ||
BC107262.2 | - | 1 | 215 | 40052516 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
