siRNA Details
Gene Symbol: Spata17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028848.3 | NP_083124.1 | 3, 4 | 458 | ||
NM_029067.1 | NP_083343.1 | 3, 4 | 458 | |||
GenBank mRNA | AY747687.1 | - | 3, 4 | 433 | ||
BC089520.1 | - | 3, 4 | 433 | 6774024 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
