siRNA Details
Gene Symbol: Tmem114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029070.2 | NP_083346.1 | 3 | 827 | ||
GenBank mRNA | AK015757.1 | - | 3 | 828 | ||
BC027071.1 | - | 3 | 800 | |||
BC147190.1 | - | 3 | 604 | 9056234 | ||
BC147191.1 | - | 3 | 604 | 9056214 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
