siRNA Details
Gene Symbol: Setmar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276356.1 | NP_001263285.1 | 3 | 586 | ||
NM_178391.4 | NP_848478.2 | 2 | 433 | |||
GenBank mRNA | AK017895.1 | - | 2 | 434 | ||
BB662956.1 | - | 2 | 434 | |||
BC045208.1 | - | 2 | 423 | 5067523 |
