siRNA Details
Gene Symbol: Rhoh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081105.1 | NP_001074574.1 | 3 | 757 | ||
GenBank mRNA | AK017885.1 | - | 3 | 743 | ||
AK137909.1 | - | 1 | 188 | |||
AK138090.1 | - | 4 | 884 | |||
AK169618.1 | - | 3 | 718 | |||
BC040409.1 | - | 1 | 132 | |||
BC094937.1 | - | 3 | 496 | 30075117 |
