siRNA Details
Gene Symbol: Yipf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205157.1 | NP_001192086.1 | 4 | 369 | ||
NM_001205158.1 | NP_001192087.1 | 4 | 359 | |||
NM_138303.2 | NP_612176.1 | 3 | 507 | |||
GenBank mRNA | AA967152.1 | - | 3 | 306 | ||
AK004959.1 | - | 3 | 293 | |||
AK033705.1 | - | 4 | 318 | |||
AK133631.1 | - | 4 | 329 | |||
BC002282.1 | - | 3 | 422 | 3497525 | ||
BC003289.1 | - | 4 | 300 | 3583087 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
