siRNA Details
Gene Symbol: Atp13a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164366.1 | NP_001157838.1 | 29 | 3566 | ||
NM_029097.2 | NP_083373.2 | 29 | 3640 | |||
GenBank mRNA | AK003623.1 | - | 13 | 1705 | ||
AK156104.1 | - | 29 | 3619 | |||
AK171305.1 | - | 29 | 3520 | |||
AK214253.1 | - | 2 | 183 | |||
BC023746.1 | - | 26 | 3942 | 5353766 | ||
BC042661.1 | - | 29 | 3585 | 4192094 |
